Learn More
Tecan Anti-HMGB1 monoclonal antibody clone DPH1.1
SDP
Supplier:  Tecan REHM902
Encompass_Preferred
anti-HMGB1 [DPH1.1] monoclonal antibody - 250?g . The mouse monoclonal IgG1 DPH1.1 was generated by injecting C57BL/6 mice with the 17-mer peptide P1 (KGKPDAAKKGVVKAEKS). Hybridomas were generated from splenocytes by standard techniques and tested by ELISA against the immunogen and full-length HMGB1. DPH1.1 mAb recognizes all mammalian HMGB1s, including human, mouse and rat. It does not recognize HMGB2. DPH1.1 mAb can be used for Western blot (WB), immunofluorescence (IHF) and immunohistochemistry (IHC). DPH1.1 mAb blocks HMGB1-elicited cell migration in trans-well migration assays. Distributed by IBL. Research Use Only.
Catalog No. 50-130-6203
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.