Learn More
Fluidigm Corp ANTI-HU CD28 -160GD
SDP
Supplier:  Fluidigm Corp 3160003B
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
CD28 is a 45 kDa disulfide-linked homodimeric type I glycoprotein. CD28 binds to CD80 and CD86 and can be expressed on T cells, NK cell subsets, and plasma cells. Expression of CD28 on T cells provides co-stimulatory signals, which is required for T cell activation for the production of various interleukins (IL-2 and IL-6 in particular).
Catalog No. 50-466-908