Learn More
Fluidigm Corp ANTI-HU CD45RO-149SM
SDP
Supplier:  Fluidigm Corp 3149001B
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
CD45RO, a splice variant of CD45, is a single chain membrane glycoprotein in the protein tyrosine phosphatase (PTP) family. It is expressed on activated memory T cells, B cell subsets, activated monocytes/macrophages, and granulocytes.
Catalog No. 50-466-840