Learn More
interferon induced transmembrane protein 3 (1-8U) (D03P), Rabbit anti-Human, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human IFITM3 protein.
Supplier: Abnova Corporation H00010410D03P
Description
Sequence: MNHTVQTFFSPVNSGQPPNYEMLKEEHEVAVLGAPHNPAPPTSTVIHIRSETSVPDHVVWSLFNTLFMNPCCLGFIAFAYSVKSRDRKMVGDVTGAQAYASTAKCLNIWALILGILMTILLIVIPVLIFQAYGSpecifications
interferon induced transmembrane protein 3 (1-8U) | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human IFITM3 protein. | |
IFITM3 | |
1-8U/IP15 | |
Rabbit | |
Affinity Purified | |
RUO | |
10410 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC006794 | |
IFITM3 | |
IFITM3 (AAH06794, 1 a.a. ∼ 133 a.a) full-length human protein. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.