Learn More
Integrin αV Mouse anti-Human, Clone: LM142, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB1978
Description
Detect Integrin αV using this Anti-Integrin αV Antibody, clone LM142 validated for use in immunoprecipitation, immunohistochemistry.Specifications
Integrin αV | |
Monoclonal | |
Unconjugated | |
ITGAV, DKFZp686A08142, MSK8, CD51, VNRA | |
100 μL | |
Human | |
Ascites |
Immunohistochemistry, Immunoprecipitation | |
LM142 | |
NM_002210.2 | |
Mouse | |
Primary | |
−20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
For Research Use Only