Learn More
Fluidigm Corp ANTI-MOUSE CD4 -145ND
SDP
Supplier:  Fluidigm Corp 3145002B
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
CD4, also known as T4, is a transmembrane glycoprotein expressed on the helper subset of T cells (thymocytes), on immature T cells in the thymus, and weakly on monocytes and dendritic cells. CD4 helps recognize antigens associated with MHC class II molecules via initiation of signal transduction and controlling of cell-cell interaction.
Catalog No. 50-466-812