Learn More
Fluidigm Corp ANTI-MOUSE CD45R/B220 -176YB
SDP
Supplier:  Fluidigm Corp 3176002B
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
CD45R, also known as B220, is the longest isoform of CD45, with a glycosylation pattern resulting in a molecular weight of 220 kDa. B cells at all developmental stages express CD45R, as do subsets of T, NK, and dendritic cells. In mice, CD45R has been historically used as a pan-B cell marker. However, CD19 offers greater specificity for the B cell lineage.
Catalog No. 50-467-013
missing translation for 'QuantityLeft'