Learn More
NPC2, Mouse, Clone: 4B9, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant NPC2.
Supplier: Abnova Corporation H00010577M01
Description
This gene encodes a protein containing a lipid recognition domain. The encoded protein may function in regulating the transport of cholesterol through the late endosomal/lysosomal system. Mutations in this gene have been associated with Niemann-Pick disease, type C2 and frontal lobe atrophy. [provided by RefSeq
Sequence: MRFLAATFLLLALSTAAQAEPVQFKDCGSVDGVIKEVNVSPCPTQPCQLSKGQSYSVNVTFTSNIQSKSSKAVVHGILMGVPVPFPIPEPDGCKSGINCPIQKDKTYSYLNKLPVKSEYPSIKLVVEWQLQDDKNQSLFCWEIPVQIVSHLSpecifications
NPC2, | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC002532 | |
NPC2 | |
NPC2 (AAH02532, 1 a.a. ∼ 151 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
ELISA, Western Blot | |
4B9 | |
Mouse monoclonal antibody raised against a full-length recombinant NPC2. | |
NPC2 | |
HE1/MGC1333/NP-C2 | |
Mouse | |
Affinity chromatography | |
RUO | |
10577 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.