Learn More
prolactin, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human PRL protein.
Supplier: Abnova Corporation H00005617D01P
Description
Sequence: MNIKGSPWKGSLLLLLVSNLLLCQSVAPLPICPGGAARCQVTLRDLFDRAVVLSHYIHNLSSEMFSEFDKRYTHGRGFITKAINSCHTSSLATPEDKEQAQQMNQKDFLSLIVSILRSWNEPLYHLVTEVRGMQEAPEAILSKAVEIEEQTKRLLEGMELIVSQVHPETKENEIYPVWSGLPSLQMADEESRLSAYYNLLHCLRRDSHKIDNYLKLLKCRIIHNNNCSpecifications
prolactin | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human PRL protein. | |
PRL | |
PRL | |
PRL (AAH15850.1, 1 a.a. ∼ 227 a.a) full-length human protein. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_000948 | |
Rabbit | |
Affinity Purified | |
RUO | |
5617 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.