Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSARL, Mouse, Clone: 4F4, Abnova™

Mouse monoclonal antibody raised against a partial recombinant PSARL.

Supplier:  Abnova Corporation H00055486M02

Catalog No. 89-002-637


missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart

Description

Description

This gene encodes a mitochondrial integral membrane protein. Following proteolytic processing of this protein, a small peptide (P-beta) is formed and translocated to the nucleus. This gene may be involved in signal transduction via regulated intramembrane proteolysis of membrane-tethered precursor proteins. Variation in this gene has been associated with increased risk for type 2 diabetes. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq

Sequence: AWRGWAQRGWGCGQAWGASVGGRSCEELTAVLTPPQLLGRRFNFFIQQKCGFRKAPRKVEPRRSDPGTSGEAYKRSALIPPVEETVFYPSPYPIRSLIK
Specifications

Specifications

PSARL
Monoclonal
Unconjugated
PBS with no preservative; pH 7.4
NM_018622
PARL
PSARL (NP_061092.3, 2 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2b κ
ELISA
4F4
Mouse monoclonal antibody raised against a partial recombinant PSARL.
PARL
PRO2207/PSARL/PSARL1/PSENIP2/RHBDS1
Mouse
Affinity chromatography
RUO
55486
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.