Learn More
arginyl aminopeptidase (aminopeptidase B), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human RNPEP protein.
Supplier: Abnova Corporation H00006051D01P
Description
Sequence: MASGEHSPGSGAARRPLHSAQAVDVASASNFRAFELLHLHLDLRAEFGPPGPGAGSRGLSGTAVLDLRCLEPEGAAELRLDSHPCLEVTAAALRRERPGSEEPPAEPVSFYTQPFSHYGQALCVSFPQPCRAAERLQVLLTYRVGEGPGVCWLAPEQTAGKKKPFVYTQGQAVLNRAFFPCFDTPAVKYKYSALIEVPDGFTAVMSASTWEKRGPNKFFFQMCQPIPSYLIALAIGDLVSAEVGPRSRVWAEPCLIDAAKEEYNGVIEEFLATGEKLFGPYVWGRYDLLFMPPSFPFGGMENPCLTFVTPCLLAGDRSLADVIIHEISHSWFGNLVTNANWGEFWLNEGFTMYAQRRISTILFGAAYTCLEAATGRALLRQHMDITGEENPLNKLRVKIEPGVDPDDTYNETPYEKGFCFVSYLAHLVGDQDQFDSFLKAYVHEFKFRSILADDFLDFYLEYFPELKKKRVDIIPGFEFDRWLNTPGWPPYLPDLSPGDSLMKPAEELAQLWAAEELDMKAIEAVAISPWKTYQLVYFLDKILQKSPLPPGNVKKLGDTYPSISNARNAELRLRWGQIVLKNDHQEDFWKVKEFLHNQGKQKYTLPLYHAMMGGSEVAQTLAKETFASTASQLHSNVVNYVQQIVAPKGSSpecifications
arginyl aminopeptidase (aminopeptidase B) | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human RNPEP protein. | |
RNPEP | |
DKFZp547H084 | |
Rabbit | |
Affinity Purified | |
RUO | |
6051 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_020216.3 | |
RNPEP | |
RNPEP (NP_064601.3, 1 a.a. ∼ 650 a.a) full-length human protein. | |
100 μg | |
Primary | |
Human | |
Antibody |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.