Learn More
SOD1, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human SOD1 protein.
Supplier: Abnova Corporation H00006647B01P
Description
The protein encoded by this gene binds copper and zinc ions and is one of two isozymes responsible for destroying free superoxide radicals in the body. The encoded isozyme is a soluble cytoplasmic protein, acting as a homodimer to convert naturally-occuring but harmful superoxide radicals to molecular oxygen and hydrogen peroxide. The other isozyme is a mitochondrial protein. Mutations in this gene have been implicated as causes of familial amyotrophic lateral sclerosis. Rare transcript variants have been reported for this gene. [provided by RefSeq
Sequence: MATKAVCVLKGDGPVQGIINFEQKESNGPVKVWGSIKGLTEGLHGFHVHEFGDNTAGCTSAGPHFNPLSRKHGGPKDEERHVGDLGNVTADKDGVADVSIEDSVISLSGDHCIIGRTLVVHEKADDLGKGGNEESTKTGNAGSRLACGVIGIAQSpecifications
SOD1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human SOD1 protein. | |
SOD1 | |
ALS/ALS1/IPOA/SOD/homodimer | |
Mouse | |
Affinity chromatography | |
RUO | |
6647 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_000454.4 | |
SOD1 | |
SOD1 (NP_000445.1, 1 a.a. ∼ 154 a.a) full-length human protein. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.