Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium/Iodine Symporter Mouse anti-Human, Clone: 2-2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB3562
Description
Anti-Sodium/Iodine Symporter Antibody, clone 2-2 detects level of Sodium/Iodine Symporter and has been published and validated for use in immunohistochemistry and western blotting.Specifications
Sodium/Iodine Symporter | |
Monoclonal | |
Unconjugated | |
NM_000453.1 | |
Mouse | |
100 μg | |
Human | |
Purified |
Immunohistochemistry, Western Blot | |
2-2 | |
Liquid. Buffer = 0.02M Phosphate buffer, 0.25M NaCl with 0.1% sodium azide. | |
SLC5A5, +I, NIS, Na, -symporter), -cotransporter) | |
Synthetic peptide corresponding to amino acids 37-54 of human NIS. | |
Primary | |
2-8°C in undiluted aliquots for up to 6 months. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction