Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Full-Finger Anti-Vibration Air Glove™ with Silicone Dots

Catalog No. 19400048 Shop All Impacto Protective Products Products
Click to view available options
Size:
This item is not returnable. View return policy
This item is not returnable. View return policy

Patented Air Glove™ technology protects hands from impact and vibration. Synthetic suede leather with silicone dots on the palm & fingers provide comfort and enhanced grip abilities.

  • Synthetic ultra suede leather provides extended wear and flexibility
  • Silicone dots on the palm and fingertips enhance grip capabilities
  • Mesh back allows ventilation and comfort
  • Durable Bubble Glove™ bladder in the palm, fingers, and thumb protects from vibration
  • Full finger style offers maximum protection and tactile feedback
  • Neoprene cuff with hook and loop closure
  • Vibration protection when using hand-held or hand guided power tools and machines

Specifications

Back Material

Mesh

Cuff Material

Neoprene

Coating Material

Silicone

Color

Black

Hand

Specific

Material

Synthetic Leather

Product Type

Anti-vibration Gloves

Size

2X-Large

Finger Style

Full

For Use With (Application)

Construction and Maintenance

Glove Cuff Style

Hook and Loop

Series

BG408

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.