Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ AntiBind™ 384 Deep Well Microplates, Conical, 120μL

Catalog No. p-6824073 Shop All DWK Life Sciences Products
Click to view available options
No. per Case:

Used in chromatography applications, long term sample storage, and bioanalytical assays. Wheaton AntiBIND 384 Deep Well Microplates, Conical, 120μL are manufactured using a new technology that modifies the polypropylene surface at the molecular level to produce a homogenous hydrophilic surface that eliminates nonspecific protein binding and ensures ultimate recovery of valuable proteins.

  • Application of Advanced Plasma Treatment technology (APT) allows for comprehensive complete hydrophilic treatment that is not compromised in presence of solvents
  • Meets the SBS and ANSI dimensions used for automation
  • Conical shaped bottoms for maximum sample recovery and efficient mixing
  • Manufactured from polypropylene resin

Specifications

Array

16 x 24

Color

Natural

For Use With (Application)

High through-put drug discovery, proteomics research, bioanalytical assays, long term sample storage, sample prep, chromatography (LC-MS)

Material

Polypropylene

No. per Case

120

Quantity

120/Cs.

Surface Treatment

AntiBIND

No. of Wells

384

Format

Standard

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.