Learn More
DWK Life Sciences Wheaton™ AntiBIND™ Deep Well Microplates
Used in chromatography applications, long term sample storage, and bioanalytical assays. Wheaton AntiBIND 96 Deep Well Microplates, Conical, are manufactured using a new technology that modifies the polypropylene surface at the molecular level to produce a homogenous hydrophilic surface that eliminates nonspecific protein binding and ensures ultimate recovery of valuable proteins.
Supplier: DWK Life Sciences W266020
Description
- Application of Advanced Plasma Treatment technology (APT) allows for comprehensive complete hydrophilic treatment that is not compromised in presence of solvents
- Meets the SBS and ANSI dimensions used for automation
- Conical shaped bottoms for maximum sample recovery and efficient mixing
- Manufactured from polypropylene resin
Specifications
0.5 mL | |
V | |
High through-put drug discovery, proteomics research, bioanalytical assays, long term sample storage, sample prep, chromatography (LC-MS) | |
20 | |
AntiBIND | |
Standard |
8 x 12 | |
Natural | |
Polypropylene | |
20/Cs. | |
96 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.