Learn More
ACROBiosystems Monoclonal Anti-DNP antibody, Human IgG1 (N297A) Isotype Control
Supplier:  ACROBiosystems DNPMB273100UG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Antibody;100UG;Human IgG1 (N297A)/kappa;Mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with DNP F(ab')2.;Monoclonal Anti-DNP (Anti-Hapten) antibody, Human IgG1 (N297A) was purified from HEK293 cell culture at ACRObiosystems, and specifically reacts with DNP (Dinitrophenyl) and DNP conjugated proteins.Dinitrophenyl (DNP) is a hapten that is not normally expressed in the target tissue. Therefore, anti-DNP antibody has no relevant specificity to a target antigen, and can be used as isotype control antibody to differentiate non-specific background signal from specific antibody signal in various in vitro and in vivo studies.
Catalog No. 50-201-9865
missing translation for 'QuantityLeft'