Learn More
ACROBiosystems Biotinylated Monoclonal Anti-DNP antibody, Human IgG1 Isotype Control
Supplier:  ACROBiosystems DNPBM2200UG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Antibody;200UG;Human IgG1/kappa;Specifically reacts with DNP (Dinitrophenyl) and DNP conjugated proteins.;Dinitrophenyl (DNP) is a hapten that is not normally expressed in the target tissue. Therefore, anti-DNP antibody has no relevant specificity to a target antigen, and can be used as isotype control antibody to differentiate non-specific background signal from specific antibody signal in various in vitro and in vivo studies. Biotinylated Monoclonal Anti-DNP antibody, Mouse IgG1 Isotype Control was purified from HEK293 cell culture. It is the Biotin labeled form ofMonoclonal Anti-DNP antibody, Human IgG1 Isotype Control (Cat. No. DNP-M2).;
Catalog No. 50-201-9413