Learn More
Fluidigm Corp Anti-Human CD122/IL-2Rb (Tu27)-170Er 100 Tests
SDP
Supplier:  Fluidigm Corp 3170004B
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
CD122 (IL-2R?) is a 70–75 kDa member of the immunoglobulin superfamily that, along with CD25 (IL-2R?) and CD132 (common ? chain), forms the receptor for the IL-2 cytokine.
Catalog No. 50-196-3277
missing translation for 'QuantityLeft'