Learn More
Fluidigm Corp Anti-Human/Mouse IL-5 (TRFK5)-143Nd 100 Tests
SDP
Supplier:  Fluidigm Corp 3143003B
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Interleukin 5 (IL-5) is a secreted protein produced by Th2-polarized T cells and mast cells. IL-5 stimulates cell division, increased antibody production, and isotype-switching in B cells.
Catalog No. 50-196-2514
missing translation for 'QuantityLeft'