Learn More
Fluidigm Corp Anti-Mouse CD1d (1B1)-162Dy 25 Tests
SDP
Supplier:  Fluidigm Corp 3162020C
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
CD1d, also known as CD1.1 and Ly-38, is a 48 kDa type I membrane glycoprotein with structural homology to harmful MHC class I molecules. It is noncovalently associated with ?2-microglobulin.
Catalog No. 50-196-3052