Learn More
Fluidigm Corp Anti-Mouse CD86 (GL1)-172Yb 25 Tests
SDP
Supplier:  Fluidigm Corp 3172016C
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
CD86, also known as B7-2, B70, and Ly-58, is expressed on activated B and T cells, macrophages, dendritic cells, and astrocytes. CD86, along with CD80, is a ligand of CD28 and CD152 (CTLA-4).
Catalog No. 50-196-3327
missing translation for 'QuantityLeft'