Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP 24534, Tocris Bioscience™

Catalog No. p-4816778 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent multi-kinase and pan-Bcr-Abl inhibitor

Potent multi-kinase and pan-Bcr-Abl inhibitor. Displays potent activity against cell lines expressing native Bcr-Abl or Bcr-AblT3151 (IC50 values are 0.37 and 2.0 nM respectively); also inhibits other Abl kinase domain mutants at nanomolar potencies. Exhibits inhibitory activity against PDGFRα, c-Src and c-Kit (IC50 values are 1.1, 5.4 and 12.5 nM respectively); potently inhibits FGFR and VEGFR family kinases. Orally active. Identified as targeting human host proteins that interact with SARS-CoV-2.

Chemical Identifiers

CAS 943319-70-8
Molecular Formula C29H27F3N6O
Molecular Weight (g/mol) 532.571
InChI Key PHXJVRSECIGDHY-UHFFFAOYSA-N
Synonym ponatinib, ponatinib ap24534, iclusig, 3-imidazo 1,2-b pyridazin-3-ylethynyl-4-methyl-n-4-4-methylpiperazin-1-yl methyl-3-trifluoromethyl phenyl benzamide, ap24534 ponatinib, iclusig tn, 3-2-imidazo 1,2-b pyridazin-3-yl ethynyl-4-methyl-n-4-4-methylpiperazin-1-yl methyl-3-trifluoromethyl phenyl benzamide, benzamide, 3-2-imidazo 1,2-b pyridazin-3-ylethynyl-4-methyl-n-4-4-methyl-1-piperazinyl methyl-3-trifluoromethyl phenyl, ponatinib usan:inn
PubChem CID 24826799
ChEBI CHEBI:78543
IUPAC Name 3-(2-imidazo[1,2-b]pyridazin-3-ylethynyl)-4-methyl-N-[4-[(4-methylpiperazin-1-yl)methyl]-3-(trifluoromethyl)phenyl]benzamide
SMILES CC1=C(C=C(C=C1)C(=O)NC2=CC(=C(C=C2)CN3CCN(CC3)C)C(F)(F)F)C#CC4=CN=C5N4N=CC=C5

Specifications

Quantity 10 mg
Formula Weight 532.56
Percent Purity >98%
Chemical Name or Material AP 24534
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.