Learn More
Biolegend APC anti-human P2RY12 [S16001E]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Supplier:  Biolegend 392114
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
APC anti-human P2RY12 [S16001E]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-272-3143
missing translation for 'QuantityLeft'