Learn More
Kamiya Biomedical Apo AII/CII/CIII Calibrator, 1x1mL Cal + 2x2mL Diluent
Supplier:  Kamiya Biomedical KAI041C
Encompass_Preferred
K-ASSAY Apo AII/CII/CIII Calibrator. 1 x 1 mL stock calibrator (lyophilized), 2 x 2 mL diluent (liquid). The calibrator and diluent are used to prepare 6 calibration points. The K-ASSAY Apo AII/CII/CIII Calibrator is intended to be used for the calibration of the K-ASSAY Apo AII, Apo CII, and Apo CIII immunoturbidimetric assays (Cat. No. KAI-003, KAI-005, and KAI-006). FOR RESEARCH USE ONLY IN THE U.S. NOT FOR DIAGNOSTIC PROCEDURES IN THE U.S.
Catalog No. 50-130-5960
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.