Learn More
Kamiya Biomedical Apo AII / CII / CIII / E Control, L1: 2 x 1mL, L2: 2 x 1mL
Supplier: Kamiya Biomedical K315C4M
K-ASSAY Apo AII / CII / CIII / E Control. Level 1: 2 x 1 mL (lyophilized), Level 2: 2 x 1 mL (lyophilized). The K-ASSAY Apo AII / CII / CIII / E Control is intended for use as an assayed quality control material for monitoring the performance of the K-ASSAY Apo AII (Cat. No. KAI-003), Apo CII (Cat. No. KAI-005), Apo CIII (Cat. Not. KAI-006), and Apo E (Cat. No. KAI-007) immunoturbidimetric assays. FOR RESEARCH USE ONLY IN THE U.S. NOT FOR DIAGNOSTIC PROCEDURES IN THE U.S.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.