Learn More
AG Scientific Inc Apomorphine HCl, 100 MG
Supplier:  AG Scientific Inc A2933100MG
Encompass_Preferred
Apomorphine Hydrochloride is the hydrochloride salt form of apomorphine; a derivative of morphine and non-ergoline dopamine agonist with high selectivity for dopamine D2, D3, D4 and D5 receptors. Apomorphine hydrochloride acts by stimulating dopamine receptors in the nigrostriatal system, hypothalamus, limbic system, pituitary gland, and blood vessels. This enhances motor function, suppresses prolactin release, and causes vasodilation and behavioral effects. Apomorphine hydrochloride is used in the treatment of Parkinson's disease and erectile dysfunction. In addition, apomorphine hydrochloride acts on the chemoreceptor trigger zone and is used as a central emetic in the treatment of drug overdose.CAS Number: 41372-20-7Molecular Weight: 312.8Chemical Formula: C17H17NO2 . HCl 1/2 H2OSolubility: May be dissolved in water (20mg/ml)Storage Temperature: Room TemperatureResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-7072
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.