Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apoptosis Activator 2, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Apoptosis activator; promotes the cytochrome c-dependent oligomerization of Apaf-1 into the mature apoptosome. Increases procaspase-9 processing and subsequent caspase-3 activation. Induces apoptosis in tumor cells (IC50 = 4 - 9 μM for leukemia cells) with weak or no effect on normal cell lines or those defective/deficient in Apaf-1, caspase-9 or caspase-3 activity (IC50 > 40 μM).
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 306.14 |
Percent Purity | >99% |
Chemical Name or Material | Apoptosis Activator 2 |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction