Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aprepitant 98.0+%, TCI America™
SDP

Catalog No. p-7118234 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
100 mg
500 mg

Chemical Identifiers

CAS 170729-80-3
Molecular Formula C23H21F7N4O3
Molecular Weight (g/mol) 534.435
MDL Number MFCD08277635
InChI Key ATALOFNDEOCMKK-OITMNORJSA-N
Synonym 5-[[(2R,3S)-2-[(1R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)-4-morpholinyl]methyl]-1,2-dihydro-3H-1,2,4-triazol-3-one
PubChem CID 6918365
ChEBI CHEBI:499361
IUPAC Name 5-[[(2R,3S)-2-[(1R)-1-[3,5-bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)morpholin-4-yl]methyl]-1,2-dihydro-1,2,4-triazol-3-one
SMILES CC(C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)OC2C(N(CCO2)CC3=NC(=O)NN3)C4=CC=C(C=C4)F

Specifications

Melting Point 252°C
Color White
Quantity 100 mg
Formula Weight 534.43
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Aprepitant
RTECSNumber XZ5480400
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.