Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APY 29, Tocris Bioscience™

Catalog No. p-4815633 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Inhibits IRE1α autophosphorylation; activates IRE1α endoribonuclease activity

Allosteric modulator of IRE1α. Inhibits IRE1α autophosphorylation (IC50 = 280 nM) and activates IRE1α ribonuclease activity.

Chemical Identifiers

CAS 1216665-49-4
Molecular Formula C17H16N8
Molecular Weight (g/mol) 332.371
InChI Key WJNBSTLIALIIEW-UHFFFAOYSA-N
Synonym apy29, apy 29, apy-29, n2-1h-benzimidazol-6-yl-n4-5-cyclopropyl-1h-pyrazol-3-yl-2,4-pyrimidinediamine, n2-1h-benzo d imidazol-6-yl-n4-5-cyclopropyl-1h-pyrazol-3-yl pyrimidine-2,4-diamine, n∼2∼-1h-benzimidazol-5-yl-n∼4∼-3-cyclopropyl-1h-pyrazol-5-yl pyrimidine-2,4-diamine, apy 29;apy-29, 2-n-1h-1,3-benzodiazol-5-yl-4-n-5-cyclopropyl-2h-pyrazol-3-yl pyrimidine-2,4-diamine, 2-n-3h-benzimidazol-5-yl-4-n-5-cyclopropyl-1h-pyrazol-3-yl pyrimidine-2,4-diamine, n2-3h-1,3-benzodiazol-5-yl-n4-5-cyclopropyl-1h-pyrazol-3-yl pyrimidine-2,4-diamine
PubChem CID 42627755
IUPAC Name 2-N-(3H-benzimidazol-5-yl)-4-N-(5-cyclopropyl-1H-pyrazol-3-yl)pyrimidine-2,4-diamine
SMILES C1CC1C2=CC(=NN2)NC3=NC(=NC=C3)NC4=CC5=C(C=C4)N=CN5

Specifications

Color White
Quantity 10 mg
Formula Weight 332.36
Percent Purity >98%
Physical Form Solid
Chemical Name or Material APY 29
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.