Learn More
OHAUS™ Aquasearcher™ Conductivity Meter AB23EC
Shop All OHAUS Products

- Equipped with a large 5 in. LCD display and six instructional menu buttons
- Features a 99 item memory for measurements and last calibration
- Choose between just the AB23EC meter with electrode holder and a full kit including AB23EC meter, electrode holder, STCON3 probe, and four conductivity standard kits (1413 μS/cm, 50 mL)
Specifications
Accuracy (Conductivity)
±1% Reading ±3 LSD
Includes
AB23EC Meter, Electrode Holder, STCON3 Probe, 4 Conductivity Standard Kits (1413 μS/cm, 50 mL)
Measurement Modes
Auto-stop, Continuous
Calibration
Linear
Calibration Points
1 Point Cell Constant
Range (Conductivity)
0.01 μS/cm to 199.9 mS/cm
Range (Salinity)
0 to 99.9 psu
Temperature Range (Metric)
0.0°C to 100.0°C
Display Type
5 in. LCD Segment Matrix
Data Points
99 Sets
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.