Learn More
Sartorius arium™ comfort I ASTM 1, 3 Combined Water purification Systems w/o UV Lamp, TOC Monitor

Use the Sartorius arium™ comfort I Combined System for producing ASTM Type 1 ultrapure water and Type 3 pure water.
Supplier: Sartorius H2OI1B
Description
- Unique cartridge kit for production of highest quality ultrapure water
- Innovative iJust technology optimizes product water quality
- Touch display with intuitive menu navigation
- Compact design with SD card slot
- Reproducible and reliable results
- arium software controls valve on concentrate outlet corresponding to measurement data for CaCO3 and CO2
- Available as bench-top system for every laboratory workstation and wall-mounted with integrated wall bracket
- Optimized, economic water consumption
- Premium product-water quality at all times
- Guaranteed longer life of downstream ultrapure water systems
Specifications
<1 CFU/L | |
8 L/hr. | |
Water Purification System | |
2°C to 30°C | |
80% | |
Wall | |
Adsorption by spherical activated carbon, catalyst, reverse osmosis, deionization, UV radiation, optional final particle and sterile filtration | |
17 in. | |
19.7 in. | |
18.7 in. | |
68.3 lb. (Operating); 50.7 lb. (Empty) | |
100/240 VAC (+/-10%) 50/60 Hz 130 VA (max.) |
120 L/hr. (Type 1), 8 L/hr. or 16 L/hr. (Type 3) | |
Type 1 and 3 | |
2.0 to 6.9 bar | |
HPLC, GC-MS, AAS, ICP-MS, Ion chromatography, TOC-Analysis, Photometry, Buffer solutions, Feed water for laboratory devices, such as autoclaves, glassware washers etc | |
UV lamp | |
<1mL | |
<1 NTU | |
43.5 cm | |
50.1 cm | |
47.6 cm | |
Approx. 31 kg (Operating); 23 kg (Empty) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.