Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aroclor 1254 Solution, 50μg/g in Transformer Oil, Ultra Scientific

Click to view available options
Quantity:
1 x 1 mL
Specifications
Specifications
Chemical Name or Material | Aroclor 1254 Solution |
Name Note | 50μg/g |
CAS | 64742-53-6, 11097-69-1 |
Quantity | 1 x 1 mL |
Concentration or Composition (by Analyte or Components) | 50μg/g |
Physical Form | Oily Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction