Learn More
AdipoGen Arsenazo III
SDP
Supplier:  AdipoGen CDXA0308G001
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Chemical. CAS 1668-00-4. Formula C22H18As2N4O14S2. MW 776.37. Synthetic Arsenazo III is a calcium-sensitive dye. It is used for the determination of Ca2 in micromolar amounts, to investigate calcium transport in permeabilized cells and determination of serum calcium. Also potentially useful for the spectrophotometric determination of uranium and thorium.
Catalog No. 50-256-1755
missing translation for 'QuantityLeft'