Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arsenic Standard For ICP, 10,000μg/mL in 5% HNO3, Assurance Grade, SPEX CertiPrep™
SDP

Catalog No. p-4426758 Shop All Spex CertiPrep Products
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Triple-leached LDPE bottle

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
As10,000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 7440-38-2, 7697-37-2, 7732-18-5
Molecular Formula As
Molecular Weight (g/mol) 74.92
MDL Number MFCD00085309
InChI Key RQNWIZPPADIBDY-UHFFFAOYSA-N
PubChem CID 5359596
ChEBI CHEBI:27563
IUPAC Name arsenic
SMILES [As]

Specifications

Packaging Triple-leached LDPE bottle
Quantity 125 mL
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Grade Assurance
Solvent or Matrix 5% HNO3
Chemical Name or Material Arsenic Standard
Hazard Category Causes skin irritation, serious eye irritation. May cause respiratory irritation
DOTInformation Hazardous
Recommended Storage Ambient

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.