Learn More
Alkali Scientific CellPro™ 2.0mL CryoLok Storage Vials, Polypropylene, with RED cap, round bottom, internal sealing ring, graduated, 5 bags of 100, Sterile, 500/each
SDP
Supplier:  Alkali Scientific C4611R
Encompass_Preferred
CellPro™ Cryovials are designed for safe storage of samples at temperatures as low as -196 °C (gas phase of liquid nitrogen only). Cryogenic Vials with externally threaded caps have a special screw cap designed with silicone O-rings to avoid the risk of sample contamination. The silicone O-rings allow for expansion and contraction in liquid nitrogen environments associated with long term, -190°C storage. CellPro™ Cryogenic Vials are manufactured from high visibility polypropylene for stability in extreme temperature conditions. CellPro™ Externally Threaded Cryogenic Vials provide a safe and secure tube for ultra-low temperature storage of valuable cell cultures, clinical hospital samples and biological samples. All CellPro Croyvials are sterilized by E-Beam and are non-pyrogenic, DNase/RNase and human DNA free.
Catalog No. 50-238-9436
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.