Learn More
CPC Scientific H-Asn-Ala-Gln-Thr-Ser-Val-Ser-Pro-Ser-Lys-Val-Ile-Leu-Pro-Arg-Gly-Gly-Ser-Val-Leu-Val-Thr-Cys-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific HIV022A
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
Asn-Ala-ICAM 1 (1-21) inhibits virus replication in HIV-1 infected HTLV-1 transformed cells (MT-2 cells). The peptide induces up to 70% inhibition of reverse transcriptase activity, up to 60% reduction of soluble virus antigen production, and inhibition of syncytium formation.ONE-LETTER SEQUENCE: NAQTSVSPSKVILPRGGSVLVTCMOLECULAR FORMULA: C99H173N29O32S1MOLECULAR WEIGHT:2313.72STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [139227-42-2], RESEARCH AREA: AntimicrobialREFERENCES: J.V.Fecondo et al., AIDS Res. Human Retroviruses, 9, 733 (1993)
Catalog No. 50-210-9571
missing translation for 'QuantityLeft'