Learn More
Pyramex™ Atoka™ Safety Eyewear
Lightweight, only 22 grams
Supplier:  Pyramex™ S9110ST
 View more versions of this product
Catalog No. 19-155-616
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Integrated nosepiece
  • Soft non-slip rubber temple tips provide non-binding fit
  • Polycarbonate lens provides 99% UVA/B protection
  • Specifications
    Specifications
    ANSI Z87.1,CSA Z94.3-07
    Clear
    Polycarbonate
    Scratch-resistant
    Clear
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    Product Certifications
    Promotions
    Promotions
    Product Content Correction
    Your input is important to us. Please complete this form to provide feedback related to the content on this product.
    Product Title
    Your feedback has been submitted: Thank you for helping us improve our website.