Learn More
Electron Microscopy Sciences Sure Shot Atomizer Model 8400CB Nickel Plated Brushed Aluminum, Chrome-Plated Brass Top 16 OZ
Supplier: Electron Microscopy Sciences 6418608
Anything that the aerosol can do, ATOMIZER can do safer. Refillable and Rechargeable. Eliminates aerosol disposal problems. All Model B sprayers come standard with Viton seals, these seals are compatible with most products. IMPORTANT: Sure Shot Sprayers are not recommended for spraying any acids. Sure Shot Sprayers will handle most chemicals, but are not designed to be used with products that are intended to etch or attack metal surfaces. Nickel Plated Brushed Aluminum and chrome-plated brass top. Comes with extra valve and 2 fine, 2 regular, 2 medium, 2 coarse and 1 pin stream nozzle.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.