Learn More
Medchemexpress LLC HY-D0183 1mg Medchemexpress, ATP-polyamine-biotin CAS:1800401-93-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYD01831MG
Encompass_Preferred
Medchemexpress, HY-D0183 1mg ATP-polyamine-biotin CAS:1800401-93-7 ATP-polyamine-biotin, the first cell-permeable ATP analogue, is an efficient kinase cosubstrate. ATP-polyamine-biotin promotes biotin labeling of kinase substrates in live cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1259
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.