Learn More
Medchemexpress LLC HY-117639 10mg Medchemexpress, AUT1 CAS:1311136-84-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11763910MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Medchemexpress, HY-117639 10mg AUT1 CAS:1311136-84-1 AUT1 is a Kv3 potassium channel modulator, with pEC50s of 5.33 and 5.31 for human recombinant Kv3.1b and Kv3.2a, respectively, exhibits 10-fold lower potency at human recombinant Kv3.3 channel (pEC50, 4.5). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9485
missing translation for 'QuantityLeft'