Learn More
Applied Biosystems™ AutoLys M TubeLifter

Once centrifugation is complete the rack of tubes is returned to the TubeLifter and the inner tube of each AutoLys M Tube is separated from the outer tube. The outer tube contains the lysate that is processed further using the MagMAX FFPE DNA/RNA Ultra Kit to recover the nucleic acids.
Specifications
For Use With (Equipment)
AutoLys TubeLifter System
Quantity
1 each
Shipping Condition
Room Temperature
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.