Learn More
Trajan Scientific and Medical™ Autosampler Syringes for Agilent GCs
Gives better performance and superior quality. Trajan Scientific and Medical Autosampler Syringes for Agilent GCs has the knowledge and expertise to manufacture an SGE syringe.
Supplier:  Trajan Scientific and Medical™ 002826
 View more versions of this product
Catalog No. 03-350-401
Description
Description
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Agilent instruments
Specifications
Specifications
Agilent Syringe
0.11 mm
Fixed Needle with GT Plunger
Cone
23 and 26 G
0.63, 0.47 mm
10 μL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.