Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversey™ Avert™ Sporicidal Disinfectant Cleaner Wipes

Catalog No. 17000043
Click to view available options
:

A ready-to-use, pre-wetted, disinfectant cleaner wipe that also kills Clostridium difficile (C. diff) spores on hard, non-porous surfaces such as those found in hospitals and other healthcare facilities.

  • 27.9 x 30.5 cm (11 x 12 in.) sporicidal wipes
  • Effective in 4 minutes against Clostridium difficile (C. diff) spores.
  • Cleans, disinfects, and deodorizes in one labor-saving step
  • Bleach-based formula means no quat-binding concerns.
  • 1 minute contact time effective against a broad spectrum of pathogens including bacteria, antibiotic resistant bacteria, viruses, and fungi
  • Meets minimum active ingredient requirements for CDC, SHEA, APIC and AORN

Specifications

Product Type

Disinfectant Wipe

Certifications/Compliance

EPA, FDA

Color

White

Container Type

Bucket

Packaging Quantity

160 Wipes/Pk., 4 Pk./Cs.

Sterility

Non-sterile

pH Range

12.5

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.