Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axitinib, Tocris Bioscience™

Catalog No. p-4816546 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent VEGFR-1, -2 and -3 inhibitor

Axitinib is a potent inhibitor of VEGFR-2, -3, and -1 (IC50 values are 0.2, 0.1-0.3, and 1.2 nM respectively). Axitinib exhibits minimal activity against a panel of ∼100 protein kinases. Axitinib inhibits angiogenesis and vascular permeability. Also a high affinity BCR-ABL1 (T315I) inhibitor (Ki = 149 pM for autophosphorylated ABL1 (T315I)). Axitinib blocks proliferation of Ba/F3 cells expressing BCR-ABL1 (T315I). Axitinib (10 mg/kg) decreases T-cell exhaustion and tumor and T-cell hypoxia and improves survival in mice in vivo. Orally available.

Chemical Identifiers

CAS 319460-85-0
Molecular Formula C22H18N4OS
Molecular Weight (g/mol) 386.473
InChI Key RITAVMQDGBJQJZ-FMIVXFBMSA-N
Synonym axitinib, inlyta, axitinibum, unii-c9lvq0yuxg, e-n-methyl-2-3-2-pyridin-2-yl vinyl-1h-indazol-6-yl thio benzamide, c9lvq0yuxg, n-methyl-2-3-1e-2-pyridin-2-yl ethenyl-1h-indazol-6-yl sulfanyl benzamide, n-methyl-2-3-e-2-pyridin-2-yl ethenyl-1h-indazol-6-yl sulfanyl benzamide, e-n-methyl-2-3-2-pyridin-2-yl vinyl-1h-indazol-6-ylthio benzamide, dsstox_cid_28975
PubChem CID 6450551
ChEBI CHEBI:66910
IUPAC Name N-methyl-2-[[3-[(E)-2-pyridin-2-ylethenyl]-1H-indazol-6-yl]sulfanyl]benzamide
SMILES CNC(=O)C1=CC=CC=C1SC2=CC3=C(C=C2)C(=NN3)C=CC4=CC=CC=N4

Specifications

Quantity 10 mg
Formula Weight 386.47
Percent Purity >99%
Chemical Name or Material Axitinib
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.