Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Axl Antibody Pair [HRP]
SDP

Rabbit Monoclonal Antibody Pair

Supplier:  Novus Biologicals™ NBP2795295PLATES

 View more versions of this product

Catalog No. NB010098


Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse Axl.
Specifications

Specifications

12.5 pg/mL
Sandwich ELISA
Antibody Pairs
Mouse
AXL oncogene, AXL receptor tyrosine kinase, AXL transforming sequence/gene, EC 2.7.10, EC 2.7.10.1, JTK11, tyrosine-protein kinase receptor UFO, UFOoncogene AXL
AXL
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Axl monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Capture Antibody: 1 mg/mL of rabbit anti-Mouse Axl monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 70 ng of recombinant Mouse Axl.
  • RUO
    Storage is content dependent.
    1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 800 pg/mL is recommended.
    12.5-800 pg/mL
    HRP
    This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse Axl.
    Sandwich ELISA
    558
    Rabbit
    5 Plates
    Cell Cycle and Replication, Phospho Specific
    Axl
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Product Certifications
    Promotions

    Promotions

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.