Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Automation Tips for Agilent™/Velocity11 VPrep™ and Bravo: 384 Tips/Rack

Comprehensive line of tips for Agilent/Velocity11 automation systems

Supplier:  Axygen™ VT38431LRS

 View more versions of this product

Catalog No. 14-223-523


Description

Description

  • Tip format: 96 tip
  • Tip volume range: 20 μL to 250 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    - With or without aerosol-resistant filters
    - Nonsterile or sterile
    - Standard or Axygen™ Maxymum Recovery™ surface
  • Compatible heads: 96LT, 8LT
  • Rack format: ANSI/SLAS
  • Rack material: Polypropylene

Order Info

Packaging:10 Racks/Pack, 5 Packs/Case

Specifications

Specifications

31 μL
Sterile
Autoclavable
Maxymum Recovery™ technology for ultra-low retention
Automation, Round
50 Racks/Cs.
Polypropylene
Non-filtered
Clear
DNase, RNase, and Pyrogen free
Rack
SBS Rack
Agilent/Velocity VPrep 16ST, Agilent/Velocity VPrep 96ST, Agilent/Velocity VPrep 384ST, Agilent Bravo 384ST
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.