Learn More
Axygen™ Automation Micropipette Tips
Axygen™ Automation Micropipette Tips are constructed of polypropylene, and feature a variety of available options to suit your needs.
$117.20 - $160.60
Specifications
Volume (Metric) 200 μL
Filtered Non-filtered
Color Clear
Rack Color Blue
Autoclavable Autoclavable
View More Specs
Products 3
Catalog Number Mfr. No. Sterility Tip Style Price Quantity & Availability  
Catalog Number Mfr. No. Sterility Tip Style Price Quantity & Availability  
14-222-912
View Documents
Axygen™
ZT200RS
Sterile Automation, Round
Pack of 960 for $117.20
Case of 5 PK for $530.50
missing translation for 'QuantityLeft'
Add to Cart
14-222-911
View Documents
Axygen™
ZT200R
Non-sterile Automation, Round
Pack of 960 for $160.60
Case of 5 PK for $714.50
missing translation for 'QuantityLeft'
Add to Cart
14-222-910
View Documents
Axygen™
ZT200LRS
Sterile Automation, Maxymum Recovery, Round N/A N/A N/A
Description
Description
  • For Tecan™/Freedom EVO&trade, Te-Mo™, Aquarius, Zymark™/Caliper, RapidPlate™, and SciClone™ Systems
  • For 96-well, Non-ANSI and ANSI/SLAS (384) format, number 5 polypropylene racks
  • Available with or without aerosol-resistant filters
  • Available sterile/non-sterile
  • Available in standard or Maxymum Recovery™ style
Order Info
96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
Specifications
Specifications
200 μL
Clear
Autoclavable
Rack
Automation Micropipette Tip
Polypropylene
Non-filtered
Blue
RNase-free, DNase-free, Nonpyrogenic
96 tips/rack, 10 racks/pack, 5 packs/case
Tecan/Freedom EVO, Te-Mo, Aquarius, Zymark/Caliper, RapidPlate, and SciClone Systems
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.