Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AZ 11645373, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent and selective human P2X7 antagonist (KB values are 5 - 7 and > 10,000 nM at hP2X7 and rP2X7 respectively) that is completely without effect at all other P2X subtypes. Inhibits BzATP-mediated calcium influx and inhibits ATP-mediated IL-1β release in vitro (KB values are 15 and 92 nM respectively).
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 463.51 |
Percent Purity | >98% |
Chemical Name or Material | AZ 11645373 |
Safety and Handling
Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction