Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azathioprine, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. p-7156948 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
10 kg
Packaging:
Amber Glass Bottle
Poly Pail

A1383, 446-86-6, C9H7N7O2S

Spectrum™ Chemical Azathioprine, USP is an immunosuppressive drug used in autoimmune diseases and organ transplants. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 446-86-6
Molecular Formula C9H7N7O2S
Molecular Weight (g/mol) 277.26
InChI Key LMEKQMALGUDUQG-UHFFFAOYSA-N
IUPAC Name 6-[(1-methyl-4-nitro-1H-imidazol-5-yl)sulfanyl]-7H-purine
SMILES CN1C=NC(=C1SC1=C2NC=NC2=NC=N1)[N+]([O-])=O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 1%
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 98 to 102%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.